Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032889.1 | NP_001028061.1 | 549 | 66 | ||
XM_015150990.2 | XP_015006476.2 | 724 | 66 | |||
GenBank mRNA | AF190943.1 | - | 549 | 66 | ||
AF190944.1 | - | 456 | 66 | |||
AJ554302.1 | - | 536 | 66 | |||
JU474751.1 | - | 677 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |