Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032865.1 | NP_001028037.1 | 286 | 82 | ||
GenBank mRNA | AF240771.1 | - | 286 | 82 | ||
JV518189.1 | - | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |