Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047135.1 | NP_001040600.1 | 637 | 85 | ||
GenBank mRNA | AF340040.1 | - | 637 | 85 | ||
JU475417.1 | - | 679 | 85 | |||
JV059710.1 | - | 687 | 85 |