Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040218.1 | NP_001035308.1 | 986 | 63 | ||
GenBank mRNA | AY635464.1 | - | 986 | 63 | ||
GU981763.1 | - | 994 | 63 | |||
JV708008.1 | - | 643 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |