Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042740.1 | NP_001036205.1 | 202 | 111 | ||
XM_028835753.1 | XP_028691586.1 | 262 | 111 | |||
GenBank mRNA | L16554.1 | - | 202 | 111 |
Gene Symbol: LOC114669819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836656.1 | XP_028692489.1 | 262 | 111 | ||
XM_028836657.1 | XP_028692490.1 | 387 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |