Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148650.2 | XP_015004136.2 | 276 | 92 | ||
GenBank mRNA | JV527813.1 | - | 221 | 92 | ||
JV625116.1 | - | 188 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |