Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087443.4 | XP_001087443.2 | 1160 | 66 | ||
GenBank mRNA | JU336690.1 | - | 494 | 66 | ||
JU477484.1 | - | 495 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |