Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028849824.1 | XP_028705657.1 | 415 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |