Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205127.2 | NP_001192056.2 | 3431 | 87 | ||
XM_015112348.2 | XP_014967834.1 | 2990 | 87 | |||
XM_015112349.2 | XP_014967835.2 | 2292 | 87 | |||
XM_015112350.2 | XP_014967836.1 | 828 | 87 | |||
GenBank mRNA | CK231727.1 | - | 259 | 87 | ||
JU471546.1 | - | 646 | 87 | |||
JV045048.1 | - | 540 | 87 | |||
JV588048.1 | - | 3648 | 87 | |||
JV622917.1 | - | 3328 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |