Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118889.2 | NP_001112361.1 | 575 | 60 | ||
NM_001346420.1 | NP_001333349.1 | 703 | 60 | |||
GenBank mRNA | CB552751.1 | - | 280 | 60 | ||
CN643832.1 | - | 476 | 60 | |||
JV046824.1 | - | 509 | 60 | |||
JV465885.1 | - | 506 | 60 | |||
JV602446.1 | - | 703 | 60 | |||
JV649198.1 | - | 574 | 60 | |||
JV667383.1 | - | 629 | 60 | |||
JV681449.1 | - | 561 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta Inventoried |