Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266537.1 | NP_001253466.1 | 204 | 115 | ||
GenBank mRNA | JU320217.1 | - | 124 | 115 | ||
JU471271.1 | - | 204 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |