Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258066.1 | NP_001244995.1 | 277 | 147 | ||
XM_028846746.1 | XP_028702579.1 | 557 | 147 | |||
XM_028846748.1 | XP_028702581.1 | 808 | 147 | |||
GenBank mRNA | JU476560.1 | - | 277 | 147 | ||
JV636289.1 | - | 271 | 147 | |||
JV719849.1 | - | 159 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |