Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092850.4 | XP_001092850.1 | 364 | 120 | ||
XM_015119320.2 | XP_014974806.1 | 239 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |