Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERTAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266761.2 | NP_001253690.1 | 1064 | 146 | ||
XM_015123952.2 | XP_014979438.1 | 1168 | 146 | |||
XM_028838378.1 | XP_028694211.1 | 1402 | 146 | |||
GenBank mRNA | JU321583.1 | - | 1070 | 146 | ||
JU473106.1 | - | 1101 | 146 | |||
JV045799.1 | - | 1067 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |