Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113829.2 | XP_014969315.2 | 940 | 130 | ||
XM_015113830.2 | XP_014969316.2 | 814 | 130 | |||
XM_015113832.2 | XP_014969318.1 | 876 | 130 | |||
XM_028833250.1 | XP_028689083.1 | 766 | 130 | |||
XM_028833251.1 | XP_028689084.1 | 835 | 130 | |||
XM_028833252.1 | XP_028689085.1 | 803 | 130 | |||
XM_028833253.1 | XP_028689086.1 | 636 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |