Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194630.1 | NP_001181559.1 | 770 | 155 | ||
GenBank mRNA | JV635842.1 | - | 711 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |