Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02840587_m1
See other DMTN GE Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
EPB49
Chromosome Location:
Chr.8: 22214344 - 22248806 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02840587_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015144913.2
XP_015000399.1
1351
143
XM_015144915.2
XP_015000401.1
1078
143
XM_015144918.2
XP_015000404.1
959
143
XM_015144919.2
XP_015000405.1
1218
143
XM_015144920.2
XP_015000406.1
1024
143
XM_015144921.2
XP_015000407.1
986
143
XM_015144923.2
XP_015000409.1
1319
143
XM_015144924.2
XP_015000410.1
914
143
XM_015144926.2
XP_015000412.1
1134
143
XM_015144929.2
XP_015000415.1
1289
143
XM_015144930.2
XP_015000416.1
1264
143
XM_015144931.2
XP_015000417.1
1082
143
XM_015144934.2
XP_015000420.1
884
143
XM_015144935.2
XP_015000421.1
949
143
XM_015144936.2
XP_015000422.1
1131
143
XM_015144937.2
XP_015000423.1
867
143
XM_015144938.2
XP_015000424.1
779
143
XM_015144940.2
XP_015000426.1
1244
143
XM_015144941.2
XP_015000427.1
839
143
XM_015144942.2
XP_015000428.1
1289
143
XM_028852432.1
XP_028708265.1
1364
143
XM_028852433.1
XP_028708266.1
1364
143
XM_028852434.1
XP_028708267.1
959
143
XM_028852435.1
XP_028708268.1
1319
143
XM_028852436.1
XP_028708269.1
750
143
XM_028852437.1
XP_028708270.1
854
143
XM_028852438.1
XP_028708271.1
858
143
XM_028852439.1
XP_028708272.1
1264
143
XM_028852440.1
XP_028708273.1
778
143
XM_028852441.1
XP_028708274.1
839
143
XM_028852442.1
XP_028708275.1
1244
143
XM_028852443.1
XP_028708276.1
843
143
XM_028852446.1
XP_028708279.1
1240
143
GenBank mRNA
JU333339.1
-
953
143
JV046423.1
-
905
143
JV046424.1
-
905
143
JV574083.1
-
968
143
JV574084.1
-
835
143
JV636019.1
-
892
143
JV636020.1
-
892
143
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0