Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848198.1 | XP_028704031.1 | 389 | 86 | ||
XM_028848200.1 | XP_028704033.1 | 398 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |