Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266711.2 | NP_001253640.1 | 538 | 80 | ||
GenBank mRNA | JU321398.1 | - | 547 | 80 | ||
JU473147.1 | - | 554 | 80 | |||
JV046863.1 | - | 538 | 80 | |||
JV637252.1 | - | 557 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |