Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099152.4 | XP_001099152.1 | 1037 | 104 | ||
XM_028852638.1 | XP_028708471.1 | 412 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |