Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194692.1 | NP_001181621.1 | 912 | 126 | ||
XM_001115831.4 | XP_001115831.1 | 891 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |