Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257733.1 | NP_001244662.1 | 771 | 87 | ||
XM_028835655.1 | XP_028691488.1 | 809 | 87 | |||
GenBank mRNA | JU321767.1 | - | 771 | 87 | ||
JU473684.1 | - | 761 | 87 | |||
JV043843.1 | - | 772 | 87 | |||
JV637526.1 | - | 781 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |