Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266865.1 | NP_001253794.1 | 199 | 156 | ||
XM_015114967.2 | XP_014970453.1 | 579 | 156 | |||
XM_028843342.1 | XP_028699175.1 | 442 | 156 | |||
GenBank mRNA | JU332907.1 | - | 199 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |