Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265692.1 | NP_001252621.1 | 227 | 120 | ||
XM_015123484.2 | XP_014978970.1 | 294 | 120 | |||
GenBank mRNA | JV598064.1 | - | 228 | 120 | ||
JV667501.1 | - | 227 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |