Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114669842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842110.1 | XP_028697943.1 | 131 | 133 | ||
GenBank mRNA | JV061198.1 | - | 57 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |