Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261752.1 | NP_001248681.1 | 1241 | 100 | ||
GenBank mRNA | JU332521.1 | - | 1251 | 100 | ||
JU473603.1 | - | 1211 | 100 | |||
JV045924.1 | - | 1241 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |