Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257882.1 | NP_001244811.1 | 162 | 101 | ||
GenBank mRNA | JU332526.1 | - | 162 | 101 | ||
JU474000.1 | - | 150 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |