Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832126.1 | XP_028687959.1 | 277 | 87 | ||
XM_028832127.1 | XP_028687960.1 | 390 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |