Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260978.1 | NP_001247907.1 | 1534 | 102 | ||
GenBank mRNA | JU320643.1 | - | 1534 | 102 | ||
JV565994.1 | - | 1605 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |