Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136529.2 | XP_014992015.1 | 1238 | 74 | ||
XM_015136532.2 | XP_014992018.1 | 1237 | 74 | |||
XM_028847479.1 | XP_028703312.1 | 4662 | 74 | |||
XM_028847480.1 | XP_028703313.1 | 1377 | 74 | |||
XM_028847481.1 | XP_028703314.1 | 4661 | 74 | |||
XM_028847483.1 | XP_028703316.1 | 4663 | 74 | |||
GenBank mRNA | JU321188.1 | - | 1209 | 74 | ||
JU321189.1 | - | 1209 | 74 | |||
JU474719.1 | - | 1199 | 74 | |||
JU474720.1 | - | 1199 | 74 | |||
JU474721.1 | - | 1199 | 74 | |||
JU474722.1 | - | 1199 | 74 | |||
JU474723.1 | - | 1199 | 74 | |||
JU474724.1 | - | 1199 | 74 | |||
JU474725.1 | - | 1199 | 74 | |||
JU474726.1 | - | 1199 | 74 | |||
JV046466.1 | - | 1209 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |