Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095989.4 | XP_001095989.1 | 357 | 120 | ||
GenBank mRNA | JV583430.1 | - | 207 | 120 | ||
JV602646.1 | - | 243 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |