Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092816.2 | XP_001092816.2 | 744 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |