Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261283.1 | NP_001248212.1 | 373 | 148 | ||
XM_015117794.2 | XP_014973280.1 | 392 | 148 | |||
XM_028834280.1 | XP_028690113.1 | 127 | 148 | |||
GenBank mRNA | JU323108.1 | - | 373 | 148 | ||
JU323213.1 | - | 373 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |