Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152609.2 | XP_015008095.2 | 928 | 144 | ||
XM_015152610.2 | XP_015008096.1 | 757 | 144 | |||
GenBank mRNA | JU488943.1 | - | 910 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |