Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135341.2 | XP_014990827.2 | 12012 | 127 | ||
XM_015135342.2 | XP_014990828.2 | 11403 | 127 | |||
XM_028846497.1 | XP_028702330.1 | 12267 | 127 | |||
XM_028846498.1 | XP_028702331.1 | 12209 | 127 | |||
XM_028846499.1 | XP_028702332.1 | 12206 | 127 | |||
XM_028846500.1 | XP_028702333.1 | 12206 | 127 | |||
XM_028846501.1 | XP_028702334.1 | 12209 | 127 | |||
XM_028846502.1 | XP_028702335.1 | 12176 | 127 | |||
XM_028846503.1 | XP_028702336.1 | 12044 | 127 | |||
XM_028846504.1 | XP_028702337.1 | 12494 | 127 | |||
XM_028846505.1 | XP_028702338.1 | 11965 | 127 | |||
XM_028846506.1 | XP_028702339.1 | 11928 | 127 | |||
XM_028846507.1 | XP_028702340.1 | 12353 | 127 | |||
GenBank mRNA | JV589493.1 | - | 278 | 127 | ||
JV589494.1 | - | 278 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |