Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257946.2 | NP_001244875.1 | 561 | 87 | ||
XM_015147984.2 | XP_015003470.2 | 821 | 87 | |||
XM_015147985.2 | XP_015003471.2 | 821 | 87 | |||
GenBank mRNA | JU335934.1 | - | 561 | 87 | ||
JU476815.1 | - | 596 | 87 | |||
JU476816.1 | - | 596 | 87 | |||
JV047943.1 | - | 596 | 87 | |||
JV721232.1 | - | 228 | 87 |