Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257946.2 | NP_001244875.1 | 1133 | 138 | ||
XM_028853483.1 | XP_028709316.1 | 679 | 138 | |||
XM_028853484.1 | XP_028709317.1 | 600 | 138 | |||
GenBank mRNA | JU335934.1 | - | 1133 | 138 | ||
JU476815.1 | - | 1168 | 138 | |||
JU476816.1 | - | 1168 | 138 | |||
JV047943.1 | - | 1168 | 138 | |||
JV721232.1 | - | 800 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |