Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257457.1 | NP_001244386.1 | 973 | 91 | ||
XM_015135530.2 | XP_014991016.1 | 1499 | 91 | |||
GenBank mRNA | JU322795.1 | - | 973 | 91 | ||
JU477233.1 | - | 977 | 91 | |||
JV048271.1 | - | 959 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |