Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257859.2 | NP_001244788.1 | 1220 | 146 | ||
XM_015136544.2 | XP_014992030.1 | 966 | 146 | |||
XM_015136545.2 | XP_014992031.1 | 959 | 146 | |||
XM_028846798.1 | XP_028702631.1 | 2126 | 146 | |||
XM_028846799.1 | XP_028702632.1 | 960 | 146 | |||
GenBank mRNA | JU333401.1 | - | 1220 | 146 | ||
JV608936.1 | - | 1235 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |