Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266830.1 | NP_001253759.1 | 757 | 135 | ||
GenBank mRNA | JU474464.1 | - | 805 | 135 | ||
JV635980.1 | - | 757 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |