Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266495.2 | NP_001253424.1 | 775 | 80 | ||
GenBank mRNA | JU326661.1 | - | 492 | 80 | ||
JV046799.1 | - | 776 | 80 | |||
JV578877.1 | - | 814 | 80 | |||
JV613671.1 | - | 815 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |