Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260538.1 | NP_001247467.1 | 539 | 162 | ||
GenBank mRNA | JV046098.1 | - | 539 | 162 |
Gene Symbol: SUPT20H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335347.1 | - | 3233 | 162 | ||
JU335348.1 | - | 3129 | 162 | |||
JU476308.1 | - | 3191 | 162 | |||
JU476309.1 | - | 3087 | 162 | |||
JV718351.1 | - | 3263 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |