Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194465.2 | NP_001181394.1 | 514 | 93 | ||
XM_028832713.1 | XP_028688546.1 | 1082 | 93 | |||
GenBank mRNA | JU321540.1 | - | 302 | 93 | ||
JU473591.1 | - | 339 | 93 | |||
JV601818.1 | - | 356 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |