Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102155.4 | XP_001102155.1 | 105 | 91 | ||
XM_028848506.1 | XP_028704339.1 | 108 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |