Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845355.1 | XP_028701188.1 | 162 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |