Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089908.4 | XP_001089908.2 | 1072 | 86 | ||
XM_015128548.2 | XP_014984034.2 | 1163 | 86 | |||
GenBank mRNA | JV643439.1 | - | 384 | 86 |
Gene Symbol: LOC114669844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842322.1 | XP_028698155.1 | 955 | 86 |
Set Membership: |
![]() ![]() ![]() |