Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122649.3 | NP_001116121.2 | 1970 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |