Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193972.2 | NP_001180901.1 | 1054 | 103 | ||
XM_028848064.1 | XP_028703897.1 | 1777 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |