Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194228.1 | NP_001181157.1 | 284 | 77 | ||
XM_028827360.1 | XP_028683193.1 | 319 | 77 | |||
XM_028827361.1 | XP_028683194.1 | 315 | 77 | |||
XM_028827362.1 | XP_028683195.1 | 315 | 77 | |||
XM_028827363.1 | XP_028683196.1 | 314 | 77 | |||
XM_028827364.1 | XP_028683197.1 | 257 | 77 | |||
GenBank mRNA | JU325710.1 | - | 219 | 77 | ||
JV046269.1 | - | 248 | 77 | |||
JV572349.1 | - | 250 | 77 | |||
JV635541.1 | - | 239 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |