Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194891.1 | NP_001181820.1 | 324 | 62 | ||
GenBank mRNA | JU322093.1 | - | 254 | 62 | ||
JV515825.1 | - | 266 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |